ContactUsLinkComponent
Sign and label printer with Wifi - AZERTY, BradyPrinter S3100
Sign and label printer with Wifi - AZERTY, BradyPrinter S3100
Catalog # BRDY149122
Supplier:  Brady
undefined
Sign and label printer with Wifi - AZERTY, BradyPrinter S3100
Catalog # BRDY149122
Supplier:  Brady
Specifications
  • Description:
    BradyPrinter S3000 sign and label printer with Wifi - AZERTY
  • Pk:
    1
Specifications
About this item
This S3100 printer is extremely easy to use and features auto-calibration and automatic label material setup
This will enable you to just walk up to the printer and start printing the signs and labels you need. Switching to another label material is easy and extremely fast thanks to the printer's 'drop-in' label materials and matching ink ribbons that can be switched out in less than 20 seconds. An intuitive user interface with touch screen offers a wide variety of sign & label design options, even when the printer is used in standalone mode.