Mop systems, HandySorb
About this item
HandySorb is a unique and practical tool to safely remove spills in a highly efficient and easy way.
Safe spill clean-up: works with inert pads or pillows that can be replaced without ever touching the absorbed fluids
Easy to use: offers 1,83 m reach when fully extended and features a no touch snap-on system for pads
Highly efficient: each pad absorbs in seconds up to 25 times its weight
2 Options Available Below
Product Details & Documents
HandySorb is a unique and practical tool to safely remove spills in a highly efficient and easy way.
Safe spill clean-up: works with inert pads or pillows that can be replaced without ever touching the absorbed fluids
Easy to use: offers 1,83 m reach when fully extended and features a no touch snap-on system for pads
Highly efficient: each pad absorbs in seconds up to 25 times its weight
Product Family Options
Product Information
- CapacityPackedAvailabilityPrice
- BRDY150597Mop system Basic universal pad refills with mop adapterMop system Basic universal pad refills with mop adapter74 L100 pads/kitLog in for availability1 * 150 ST341,00 €
- BRDY150598Mop system 'No-Touch' universal pads