ContactUsLinkComponent
Absorbent pillows, ALLWICK Universal
Absorbent pillows, ALLWICK Universal
Catalog # BRDY813792
Supplier:  Brady
Absorbent pillows, ALLWICK Universal
Catalog # BRDY813792
Supplier:  Brady
Specifications
  • Capacity:
    104 L
  • L×W:
    430×480 mm
  • Packed:
    16 pieces/box
  • Pk:
    16
Specifications
About this item
These absorbent pillows absorb spills and leaking fluids in tight spaces and under persistent drip sources.
  • Absorbs a variety of liquids including water, coolants, solvents, oils, antifreeze, etc
  • Ideal under machines or in tight spaces