Absorbent socs, Allwik Universal
: Brady
These Allwik-Socs are used for absorbing leaks and spills around machines.
- Absorbs a variety of industrial liquids including aggressive chemicals
- Flexible design allows the soc to fit around any machine, corner or curve
- Filled with universal meltblown polypropylene
Frequently Bought Together