Avantor® Hichrom HI-200, GC Columns
: Avantor
Unique selectivity Trifluoropropyl Methyl Polysiloxane stationary phase.
- Phase composition: Crossbonded Trifluoropropyl Methyl Polysiloxane
- Long lifetime
Max temperature limits may change depending on the film thickness.
- Catalog No:
- HI32-25-100-105
- HI32-10-020-15
- HI32-25-100-10
- HI32-05-005-5
- HI32-25-025-30
- HI32-10-010-10
- HI32-25-050-10
- HI32-05-005-2
- HI32-05-005-2.5
- HI32-25-050-15
- HI32-10-020-10
- HI32-18-018-10
- HI32-25-050-105
- HI32-18-018-50
- HI32-32-150-105
- HI32-18-018-40
- HI32-18-020-10
- HI32-25-050-60
- HI32-18-020-40
- HI32-32-150-60
- HI32-53-025-15
- HI32-53-010-30
- HI32-32-100-60
- HI32-20-100-40
- HI32-20-100-50
- HI32-25-010-30
- HI32-25-010-15
- HI32-25-025-105
- HI32-32-050-50
- HI32-32-100-10
- HI32-32-100-15
- HI32-32-100-105
- HI32-32-100-50
- HI32-32-100-30
- HI32-32-050-105
- HI32-32-050-10
- HI32-20-020-20
- HI32-20-020-50
- HI32-20-020-40
- HI32-20-040-10
- HI32-53-100-60
- HI32-53-150-15
- HI32-20-040-50
- HI32-32-010-50
- HI32-53-150-50
- HI32-32-025-105
- HI32-32-025-10
- HI32-53-150-60
- HI32-32-025-60
- HI32-32-025-50
- HI32-25-100-30
- HI32-15-015-15
- HI32-10-040-5
- HI32-25-100-50
- HI32-15-015-10
- HI32-32-010-105
- HI32-20-010-20
- HI32-15-015-30
- HI32-20-010-10
- HI32-32-010-15
- HI32-20-010-50
- HI32-20-010-40
- HI32-53-050-10
- HI32-53-050-50
- HI32-53-050-30
- Phase name:
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- HI-200
- Phase composition:
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Trifluoropropyl Methyl Polysiloxane
- Polarity:
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- USP code:
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
- G6
Frequently Bought Together