Avantor® Hichrom HI-1, GC Columns
: Avantor
New Product
Really versatile general purpose apolar phase for a wide range of applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 is a crossbonded phase and assure a long lifetime even under high temperature limits.
- Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- Long lifetime
Max temperature limits may change depending on the film thickness.
- Catalog No:
- HI01-53-050-10
- HI01-32-025-60
- HI01-32-025-50
- HI01-53-050-50
- HI01-18-020-10
- HI01-53-025-15
- HI01-18-040-20
- HI01-32-025-15
- HI01-15-015-30
- HI01-18-020-40
- HI01-32-025-105
- HI01-18-040-60
- HIGCHI01-32-120-20
- HIGCHI01-32-132-25
- HIGCHI01-32-120-50
- HI01-53-700-15
- HI01-53-500-75
- HI01-32-500-30
- HI01-32-010-105
- HI01-32-010-10
- HI01-53-010-10
- HI01-25-100-60
- HI01-32-500-60
- HI01-25-100-10
- HI01-25-025-105
- HI01-32-500-105
- HIGCHI01-32-120-25
- HI01-32-400-30
- HI01-25-050-30
- HI01-25-010-50
- HIGCHI015308810MTX
- HI01-10-010-15
- HIGCHI015300905MTX
- HI01-25-025-30
- HI01-25-025-15
- HI01-25-100-105
- HI01-18-018-20
- HI01-10-020-5
- HI01-10-020-10
- HI01-15-015-10
- HIGCHI01-32-052-25
- HI01-10-040-10
- HI01-18-018-50
- HI01-10-010-10
- HI01-53-300-75
- HI01-53-300-50
- HI01-25-050-15
- HI01-25-050-105
- HI01-32-300-30
- HI01-53-500-50
- HI01-32-300-15
- HIGCHI01-53-015-15
- HI01-32-300-105
- HI01-32-150-30
- HI01-20-040-50
- HI01-20-040-40
- HI01-32-150-105
- HI01-53-300-10
- HI01-20-100-50
- HI01-20-100-40
- HI01-20-100-10
- HIGCHI01-25-025-25
- HI01-05-005-2.5
- HIGCHI01-25-030-30
- HIGCHI01-53-500-25
- HIGCHI01-53-265-30
- HI01-53-100-60
- HI01-32-100-105
- HI01-53-150-30
- HI01-53-150-75
- HI01-32-100-50
- HI01-32-100-30
- HI01-32-100-15
- HI01-53-150-50
- HI01-20-010-50
- HI01-32-050-15
- HI01-53-100-50
- HI01-32-050-105
- HI01-20-020-20
- HI01-32-100-10
- HI01-32-050-60
- HI01-25-025-60G5
- HI01-20-040-10
- HI01-53-100-30G5
- HI01-25-025-15G5
- HI01-25-025-30G5
- Phase name:
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- HI-1
- Phase composition:
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- USP code:
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
Frequently Bought Together