Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Inks
Ink cartridge for J2000 printer, CMY
Ink cartridge for J2000 printer, CMY
Ink cartridge for J2000 printer, CMY
Catalog # BRDY150154
Supplier:
Brady
Expand image
Ink cartridge for J2000 printer, CMY
Catalog # BRDY150154
Supplier:
Brady
Specifications
For Use With:
J2000 printer
Colour:
Cyan/magenta/yellow
Description:
Size(L×W×H): 60,32×28,57×41,27 mm
Pk:
1
Specifications
About this item
Product Details
Product Details