ContactUsLinkComponent
Chemical sorbent socs
Chemical sorbent socs
Catalog # BRDY813850
Supplier:  Brady
Chemical sorbent socs
Catalog # BRDY813850
Supplier:  Brady
Specifications
  • Description:
    Chemical sorbent socs
  • Absorbency:
    43 L
  • Pk:
    12
  • Ø×L:
    76×1220 mm
Specifications
About this item
These chemical sorbent socs filled with chemical meltblown polypropylene that absorbs chemicals and hazardous liquids.
  • Strong wicking action
  • Green colour
  • Ideal sorbent for containing and absorbing leaks and spills
Absorption begins instantly after contact with the spilled liquid, no need to turn the soc. Flexible design allows the soc to fit around any machine, corner or curve.