Spill kits, acid, base and battery acid
Supplier: Brady
Certificates
About this item
These acid spill kit, base spill kit and battery acid spill kit safely remove acid, base and battery acid spills respectively.
- Highly absorbing pads to wipe down equipment and remove spills
- SOC's to prevent spills from spreading on the workfloor
- Neutralising polymers to neutralise acids or bases
- Gloves and glasses to protect employees
- Waste disposal bag to contain all used equipment
These kits each contain a selection of solutions that enable users to combat small to medium spills.
Delivery: Each spill kit is included with practical, visual instructions for optimal product use.
Frequently Bought Together