Labels for BBP33 printers, type B-434
Supplier: Brady
Certificates
About this item
Metallised polyester (B-434) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
- Glossy, light gray finish gives labels a rating and serial plate appearance
- Service temperature from –40 °C to +90 °C
Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces.
Certifications: UL approved, CSA accepted.