Labels for BBP33 printers, type B-430
: Brady
Glossy metallised permanent polyester (B-435) is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
- Durable permanent acrylic adhesive withstands numerous solvents and variable temperatures
- Thermal transfer printable
- Service temperature of –40 to +90 °C
: UL approved, CSA accepted.
Frequently Bought Together