Pyronin G, high purity biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-32-0
:
Formula:C₁₇H₁₉ClN₂O
MDL Number:MFCD00011725
:302.8 g/mol
Merck Index:12,08190
Melting Pt:250…260 °C
EINECS:202-147-8
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥50 %
- UV Lambda max:546 to 551 nm (in 50% ethanol)
- Absorption Ratio P-15/P+15:1.23 to 1.7
Specification Test Results
Frequently Bought Together