Chemical resistant overalls, AlphaTec® 1800 Ts PLUS, Model 122
: Ansell
Lightweight, breathable microporous laminate technology, offering Type 4/5/6 protection.
- Protects against low hazard liquid spray and fine particulates
- Comfortable moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5 and AATCC 76
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Coveralls feature a 3-piece hood, single zipper with re-sealable storm flap, finger loops, and chin strap. Tunnelled elasticated wrists and hood. Attached boots with anti-slip soles. Garments are manufactured from non-woven microporous polyethylene laminate. Stitched and taped seams.
Recommended for cleanroom cleaning and preparing, body painting including surface inspection, production line support and maintenance, plant and machine cleaning/maintenance, fibre glass resin application, and powder handling.
: Type 4-B; Type 5-B, Type 6-B, EN 1073-2; EN 14126; EN 1149-5; Category III
: Individually packaged; size S-XL 40 pieces/carton, size 2XL-5XL 30 piece/carton.
- Catalog No:
- ANSEWH18T-00122-06
- ANSEWH18T-00122-04
- ANSEWH18T-00122-05
- ANSEWH18T-00122-07
- ANSEWH18T-00122-08
- ANSEWH18T-00122-09
- ANSEWH18T-00122-02
- ANSEWH18T-00122-03
- Model:
- 122
- 122
- 122
- 122
- 122
- 122
- 122
- 122
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
Frequently Bought Together