
Iron(III) acetylacetonate for synthesis, Sigma-Aldrich®
Catalog # 8.03912.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:1.7 g/l (20 °C)
- Melting Point:180 - 183 °C
- Molar Mass:353.17 g/mol
- Bulk Density:500 kg/m3
- Vapor Pressure:2.6 hPa (110 °C)
- Density:0.8 g/cm3
- Synonyms:Tris(acetylacetonato)iron(III), Tris(2,4-pentanedionato)iron(III)
- Hill Formula:C15H21FeO6
Specification Test Results
About this item
:
:353.18 g/mol
Boiling Pt:100 °C (9.7513 mmHg)
Melting Pt:180…183 °C
Density:5.24 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:237-853-5
:GHS07
CAS Number:14024-18-1
Formula:C₁₅H₂₁FeO₆
MDL Number:MFCD00000020