
1-Acetylnaphthalene 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:941-98-0
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00004013
:170.21 g/mol
Boiling Pt:302 °C (760 mmHg)
Melting Pt:12 °C
Density:1.119 g/cm³
Flash Pt:>110 °C
EINECS:213-384-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6270 to 1.6290 (20°C, 589 nm)
- GC:≥94 %
- Composition:≤2 % (CoCl2) Typical of fatty acids:
Specification Test Results
Frequently Bought Together