N,N-Dimethyl-1-naphthylamine ≥98% (by GC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:86-56-6
:
Formula:C₁₂H₁₃N
MDL Number:MFCD00003919
:171.24 g/mol
Merck Index:12,03301
Boiling Pt:139...140 °C/13 mm Hg (lit.)
Density:1.042 g/ml (25 °C) (lit.)
Flash Pt:113 °C (closed cup)
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:201-682-4
:GHS07
:GHS09
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (colour):Faint yellow to dark yellow
- Appearance (form):Liquid
- Titration (nt) HClO₄ 0.1M:98.0 - 102.0 %
- Purity (GC area %):≥ 98.0 %
- Infrared spectrum:Conforms to structure
- Recommended retest period:3 years
Specification Test Results
Frequently Bought Together