

Triphenylphosphine
Catalog # ICNA0520998780
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:262.29 g/mol
Merck Index:12,09873
Boiling Pt:377 °C (1013 hPa)
Melting Pt:79 °C
Density:1.194 g/cm³ (20 °C)
Flash Pt:180 °C
EINECS:210-036-0
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:603-35-0
Formula:C₁₈H₁₅P
MDL Number:MFCD00003043