Tetradecyltrimethylammonium bromide, Sigma-Aldrich®
: Merck
:1119-97-7
:8044-71-1
:C₁₇H₃₈BrN
:MFCD00011770
:336.394 g/mol
:12,06419
:245…250 °C
:Ambient
:214-291-9
:3261
:8,II
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together