Order Entry
Austria
ContactUsLinkComponent
Tetradecyltrimethylammonium bromide
Tetradecyltrimethylammonium bromide
Catalog # MOLEM79329880
Supplier:  Molekula
CAS Number:  
Tetradecyltrimethylammonium bromide
Catalog # MOLEM79329880
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :336.394 g/mol
Merck Index:12,06419
Melting Pt:245…250 °C
Storage Temperature:Ambient
EINECS:214-291-9
UN:3261
ADR:8,II
 :GHS05
CAS Number:1119-97-7
 :8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770