Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Aluminium sulphate tetradecahydrate, GPR RECTAPUR®
Catalog # 21070.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Aluminium sulphate tetradecahydrate, GPR RECTAPUR®
Catalog # 21070.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (calculated as Al2O3):
    16.5 - 17.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Not precipitated by NH4OH (as SO4):
    Max. 0.3 %
  • Cl (Chloride):
    Max. 100 ppm
  • Fe (Iron):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :594.37 g/mol
EINECS:233-135-0
 :GHS05
CAS Number:16828-12-9
Formula:Al₂(SO₄)₃·14H₂O