Calcium titanate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12049-50-2
:
Formula:CaO₃Ti
MDL Number:MFCD00015981
:135.96 g/mol
Melting Pt:1975 °C
Density:4.10
Storage Temperature:Ambient
EINECS:234-988-1
Frequently Bought Together