Magnesium titanate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12032-30-3
:
Formula:MgO₃Ti
MDL Number:MFCD00016198
Melting Pt:1610 °C
Density:3.36
:120.18 g/mol
Storage Temperature:Ambient
EINECS:234-766-4
Related Information
Frequently Bought Together