Order Entry
Austria
ContactUsLinkComponent
 
Disodium hydrogen phosphate, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
Disodium hydrogen phosphate, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Assay:
    min. 99.0 %
  • Chloride (Cl):
    max. 0.002 %
  • Iron (Fe):
    max. 0.002 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.01 %
  • Loss on Drying at 105°C:
    max. 0.2 %
  • pH of 5% Solution at 25°C:
    8.7 - 9.3
  • Sulfate (SO4):
    max. 0.005 %

 

 

Frequently Bought Together