Order Entry
Austria
Orders LinkContactUsLinkComponent
Sodium chloride

Sodium chloride

Glass bottle for solids, 500 g
Catalog # OXOILP0005B
Sodium chloride
Catalog # OXOILP0005B
CAS Number:  7647-14-5
Sodium chloride

Glass bottle for solids, 500 g

CAS Number:  7647-14-5
Price
43,60 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Saline, NaCl, Sea salt, Halite
  • CAS Number:7647-14-5
  • Formula:NaCl
  • MDL Number:MFCD00003477
  • Molecular Weight:58.44 g/mol
  • Merck Index:14,08599
  • Boiling Pt:1413 °C (1013 hPa)
  • Melting Pt:801 °C
  • Density:2.16 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-598-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications