
Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
:7647-14-5
:
:NaCl
:MFCD00003477
:58.44 g/mol
:14,08599
:1413 °C (1013 hPa)
:801 °C
:2.16 g/cm³ (20 °C)
:231-598-3
:GHS07
:Ambient
- USP - Assay (NaCl)(USP) (dried basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Acidity or Alkalinity:Passes Test
- USP - Iodide (I):Passes Test
- USP - Barium (Ba):Passes Test
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Sulfate (SO₄):≤200 ppm
- USP - Phosphate (PO₄):≤25 ppm
- USP - Iron (Fe):≤2 ppm
- USP - Bromide (Br):≤100 ppm
- USP - Appearance of Solution:Passes Test
- USP - Ferrocyanide:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Potassium (K):≤500 ppm
- FCC - Assay (as NaCl) (FCC) (ignited basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤1 ppm
- FCC - Calcium and Magnesium:≤0.35 %
- FCC - Heavy Metals (as Pb):≤2 ppm
- FCC - Loss on Drying at 105°C:≤0.5 %
- Solubility (10 in 50):Passes Test
Frequently Bought Together