Industrial gloves, HyFlex® 11-515
Supplier: Ansell
Certificates
About this item
This Kevlar®/stainless steel/Spandex glove with foam nitrile palm coating combines high visibility with superior cut resistance and durability.
- Ability to handle sharp parts and knives with confidence, increasing worker output and security in light oil or dry applications
- Provides ANSI Level 4 abrasion resistance for longer life
- New coating technology provides increased wear in high stress areas
- Silicone-free and latex-free for worker and product protection
- Ergonomically designed so users are more likely to wear the glove
- Helps keep workers visible in low light situations thus reducing the opportunities for hand injuries to occur
- Extended cuff provides additional cut protection in the highly sensitive wrist area
The HyFlex® 11-515 glove has several outstanding features, including INTERCEPT TECHNOLOGY® yarn developed by Ansell and DuPont™, which incorporates Kevlar® Stretch Armor Technology.
Recommended for: Press shop operations; handling metal sheets & panels; inspection, selecting, checking parts; processing sheets, coils, tubes; cutting glasses, sheets, panels
Certifications: EN 388:2016 (4X42E); Cat. II
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-515/10
- ANSE11-515/11
- ANSE11-515/9
- ANSE11-515/7
- ANSE11-515/6
- ANSE11-515/8
- Model:
- 11-515
- 11-515
- 11-515
- 11-515
- 11-515
- 11-515
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Kevlar®/steel
- Kevlar®/steel
- Kevlar®/steel
- Kevlar®/steel
- Kevlar®/steel
- Kevlar®/steel
- Finish:
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- Anti-static:
- +
- +
- +
- +
- +
- +