Thick butyl gloves provide the best resistance to the most aggressive chemicals, without compromising on dexterity or comfort.
- Flexible and excellent dexterity
- Unflocked butyl glove
- Longer gauntlet style cuff for extra protection
- Beaded cuff for tear resistance and easy donning
- Rough finish on the hand for safer handling
Designed for comfort, with a natural, curved ergonomic shape and soft feel. Excellent protection against gas, ozone in particular.
Recommended for: Chemical handling, charging and blending raw materials, equipment repair & maintenance, opening and draining pumps valves or lines.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- ANSE38-560/11
- ANSE38-560/10
- ANSE38-560/9
- ANSE38-560/7
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Rough finish
- Rough finish
- Rough finish
- Rough finish
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 (ABIKPT); EN ISO 374-5 VIRUS; EN 388:2016 (1020A)
- EN ISO 374-1:2016 (ABIKPT); EN ISO 374-5 VIRUS; EN 388:2016 (1020A)
- EN ISO 374-1:2016 (ABIKPT); EN ISO 374-5 VIRUS; EN 388:2016 (1020A)
- EN ISO 374-1:2016 (ABIKPT); EN ISO 374-5 VIRUS; EN 388:2016 (1020A)
- Anti-static:
- +
- +
- +
- +