

Phenothiazine
Catalog # MOLE90028302
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:199.28 g/mol
Merck Index:12,07404
Boiling Pt:371 °C (1013 hPa)
Melting Pt:182…185 °C
Density:1.34 g/cm³ (20 °C)
Flash Pt:202 °C
EINECS:202-196-5
:GHS07
:GHS09
CAS Number:92-84-2
Formula:C₁₂H₉NS
MDL Number:MFCD00005015