Phenothiazine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:92-84-2
:
Formula:C₁₂H₉NS
MDL Number:MFCD00005015
:199.28 g/mol
Merck Index:12,07404
Boiling Pt:371 °C (1013 hPa)
Melting Pt:182…185 °C
Density:1.34 g/cm³ (20 °C)
EINECS:202-196-5
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:202 °C
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:- 0.001 g/l (20 °C)
- Melting Point:182 - 185 °C
- Molar Mass:199.26 g/mol
- Bulk Density:650 kg/m3
- Boiling Point:381 °C (1011 hPa)
- Vapor Pressure:- 0.001 hPa (30 °C)
- Density:1.34 g/cm3 (20 °C)
- pH:6 (10 g/l, H2O, 20 °C) (aqueous suspension)
- Ignition Point:470 °C
- Hill Formula:C12H9NS
Specification Test Results
Related Information
Frequently Bought Together