Tetrachloro-p-benzoquinone
EHERC11130000
:
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
:
:245.88 g/mol
:13,02088
:270 °C (1013 hPa)
:290…293 °C
:1.97 g/cm³ (20 °C)
:>100 °C
:204-274-4
:GHS07
:GHS09
:Ambient
:118-75-2
:C₆Cl₄O₂
:MFCD00001594