2,6-Di-tert-butyl-p-cresol
Catalog # KRAE17-100.9025
Supplier: KRAEMER MARTIN
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4
CAS Number:128-37-0
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644