Chloramine-T (sodium salt) trihydrate 98%
: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
:7080-50-4
:
:CH₃C₆H₄SO₂NClNa·3H₂O
:MFCD00149066
:14,2075
:167…170 °C
:1.43 g/cm³ (20 °C)
:192 °C (377 °F)
:Ambient
:204-854-7
:8,III
:281.69 g/mol
:3263
:GHS08
:GHS05
:GHS07
- Environmentally Preferable:
Frequently Bought Together