4'-Hydroxy-3'-methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:498-02-2
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00008747
:166.18 g/mol
Merck Index:13,00746
Boiling Pt:295 °C (1013 hPa)
Melting Pt:115 °C
:M-Clarity™ quality level = MQ300
EINECS:207-854-5
Frequently Bought Together