Order Entry
Austria
ContactUsLinkComponent
4'-Hydroxy-3'-methoxyacetophenone
4'-Hydroxy-3'-methoxyacetophenone
Catalog # ROTH7884.1
Supplier:  Roth Carl
CAS Number:  
4'-Hydroxy-3'-methoxyacetophenone
Catalog # ROTH7884.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :166.18 g/mol
Merck Index:13,00746
Boiling Pt:295 °C (1013 hPa)
Melting Pt:115 °C
EINECS:207-854-5
CAS Number:498-02-2
Formula:C₉H₁₀O₃
MDL Number:MFCD00008747