2-Methoxy-4-nitrophenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3251-56-7
:
Formula:C₇H₇NO₄
MDL Number:MFCD00012143
:169.14 g/mol
Melting Pt:99…104 °C
EINECS:221-839-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Melting point:99 to 104 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together