Order Entry
Austria
ContactUsLinkComponent
Neutral red solution, Sigma-Aldrich®
Neutral red solution, Sigma-Aldrich®
Catalog # N6264-50ML
CAS Number:  
undefined
Neutral red solution, Sigma-Aldrich®
Catalog # N6264-50ML
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :288.78 g/mol
Storage Temperature:Ambient
CAS Number:553-24-2
Formula:C₁₅H₁₇ClN₄
MDL Number:MFCD00012651