Oil red O 0.5% in isopropanol (IPA), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1320-06-5
:
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898
:408.51 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:12.0 °C (closed cup)
:GHS02
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance
dark red, sediment free:Conforms - Absorbance
diluted 0.25ml to 50 ml with Isopropyl alcohol:0.20 - 0.50 - Lambda Max:510 - 517 nm
Specification Test Results
Frequently Bought Together