MES (β-(N-Morpholino)ethanesulphonic acid), anhydrous ≥99%, high purity suitable for buffer solutions
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:4432-31-9
:
Formula:C₆H₁₃NO₄S
MDL Number:MFCD00006181
:195.24 g/mol
Merck Index:13,05929
Melting Pt:316 °C
Storage Temperature:Ambient
EINECS:224-632-3
Specification Test Results
- Abs.@290 nm (20%, Water) (w/w):<0.05
- Heavy Metals:≤10 ppm
- pKa (20 °C):5.9 – 6.3
- Purity:≥99%
- Solubility (10%, Water):Pass
- Water (KF):≤1%
Specification Test Results
Related Information
Frequently Bought Together