
Dimethyl glyoxime ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Catalog # 1.03062.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:0.6 g/l (20 °C)
- Melting Point:239 - 243 °C (decomposition)
- Molar Mass:116.11 g/mol
- Bulk Density:500 kg/m3
- Hazard Class:4.1
- Hazard Class Description:Flammable solid
Specification Test Results
About this item
:
:116.12 g/mol
Merck Index:13,03272
Boiling Pt:53 °C (8 mmHg)
Melting Pt:240…241 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-420-1
UN:1325
ADR:4.1,III
:M-Clarity™ quality level = MQ100
:GHS02
CAS Number:95-45-4
Formula:CH₃C(=NOH)C(=NOH)CH₃
MDL Number:MFCD00002117