

Methyl carbamate 99%
:598-55-0
:
:C₂H₅NO₂
:MFCD00007964
:75.07 g/mol
:12,06115
:176…177 °C (760 mmHg)
:54…56 °C
:1,1361 g/cm³ (56 °C)
:93 °C
:Ambient
:209-939-2
:GHS08
:GHS07
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Melting point:53 to 56 °C
- GC:≥98.5 %
Frequently Bought Together