Specifications
- Pk:200 mg
- Seq:2530
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 94.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
Synonyms: (1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene)(chloro)gold, 1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride
:
:621.02 g/mol
Storage Temperature:Refrigerator
CAS Number:852445-83-1
Formula:C₂₇H₃₆AuClN₂
MDL Number:MFCD09839143