Order Entry
Austria
ContactUsLinkComponent
2-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
2-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
Catalog # TCIAC0754-25G
Supplier:  TCI
CAS Number:  
2-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
Catalog # TCIAC0754-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Purity (Potassium iodate Method):
    min. 95.0 %

Specification Test Results

About this item

 :
 :383.24 g/mol
CAS Number:198279-94-6
Formula:C₁₂H₁₄Cl₂N₄.H₂SO₄
MDL Number:MFCD00060255