Nɑ-Carbobenzoxy-D-asparagine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Nα-Benzyloxycarbonyl-D-asparagine, Nɑ-Cbz-D-asparagine, Z-D-Asn-OH, Cbz-D-Asn-OH, N-Cbz-D-Asparagine
CAS Number:4474-86-6
:
Formula:C₁₂H₁₄N₂O₅
MDL Number:MFCD00065696
:266.25 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Specific rotation [a]20/D:-5.0 to -8.0 deg (C=1.6, AcOH)
- Solubility in hot Water:almost transparency
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together