Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
RAL 555 Kit
RAL 555 Kit Fast Acting Variation of May-Grünwald Giemsa Staining
RAL 555 Kit Fast Acting Variation of May-Grünwald Giemsa Staining
RAL 555 Kit Fast Acting Variation of May-Grünwald Giemsa Staining
Catalog # 720-0351
Supplier:
RAL
CAS Number:
Certificates
Expand image
RAL 555 Kit Fast Acting Variation of May-Grünwald Giemsa Staining
Catalog # 720-0351
Supplier:
RAL
CAS Number:
Certificates
Specifications
Pk:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
Documentation
:
:
1
Product Details
Documentation