(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥97.0% (by GC, titration analysis)
Catalog # TCIAB3035-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Optical purity (LC):min. 98.0 ee%
- Purity (Iodometric Titration):min. 97.0 %
- Melting point:102.0 to 106.0 deg-C
Specification Test Results
About this item
Synonyms: (R,R)-(-)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane, (-)-DIPAMP, (R,R)-DIPAMP, (R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], 1,2-Bis((R)-(2-methoxyphenyl)(phenyl)phosphino)ethane, (1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
:
:458.48 g/mol
:GHS07
CAS Number:55739-58-7
Formula:C₂₈H₂₈O₂P₂
MDL Number:MFCD05863546