1-(2-Bromoisovaleryl)urea ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2842-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:151.0 to 155.0 deg-C
Specification Test Results
About this item
Synonyms: Bromovalerylurea, 2-Bromo-N-carbamoyl-3-methylbutanamide
:
:223.07 g/mol
Merck Index:14,01397
EINECS:207-825-7
:GHS07
CAS Number:496-67-3
Formula:C₆H₁₁BrN₂O₂
MDL Number:MFCD00047873