Order Entry
Austria
ContactUsLinkComponent
 
3,9-Bis[2-(3,5-diamino-2,4,6-triazaphenyl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1724-25G
Supplier:  TCI
CAS Number:  
3,9-Bis[2-(3,5-diamino-2,4,6-triazaphenyl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1724-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 10.0 %

Specification Test Results

About this item

Synonyms: CTU Guanamine
 :
 :434.46 g/mol
EINECS:245-064-2
 :GHS07
CAS Number:22535-90-6
Formula:C₁₇H₂₆N₁₀O₄
MDL Number:MFCD00191395