Bis(dithiobenzil)nickel(II) ≥95.0% (by titrimetric analysis)
Catalog # TCIAB1350-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark green to Dark blue to Black powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
- Lambda max.:853.0 to 857.0 nm (Toluene)
- Absorbance (E1%1cm):min. 450 (Toruene, 853.0 to 857.0 nm)
Specification Test Results
About this item
:
:543.40 g/mol
EINECS:249-353-4
:GHS08
CAS Number:28984-20-5
Formula:C₂₈H₂₀NiS₄
MDL Number:MFCD00059034