Order Entry
Austria
ContactUsLinkComponent
Triacetin (glycerol triacetate) ≥99%, TECHNICAL
Triacetin (glycerol triacetate) ≥99%, TECHNICAL
Catalog # 24391.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Triacetin (glycerol triacetate) ≥99%, TECHNICAL
Catalog # 24391.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Refractive index (20°C):
    1.429 - 1.434
  • Water:
    Max. 0.2 %

Specification Test Results

About this item

 :
 :218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716