Iron(III) fluoride trihydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15469-38-2
:
Formula:F₃Fe·3H₂O
MDL Number:MFCD00149718
Density:2.3 g/cm³ (20 °C)
Molecular Weight:166.89 g/mol
Storage Temperature:Ambient
EINECS:232-002-4
UN:3260
ADR:8 II
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together